Thermo Scientific Chemicals

Benzyl chloroacetate, 98%, Thermo Scientific Chemicals

Catalog number: 336160050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

Benzyl chloroacetate, 98%, Thermo Scientific Chemicals

Catalog number: 336160050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 336160050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-18-1
IUPAC Name
benzyl 2-chloroacetate
Molecular Formula
C9H9ClO2
InChI Key
SOGXBRHOWDEKQB-UHFFFAOYSA-N
SMILES
ClCC(=O)OCC1=CC=CC=C1
Appearance (Form)
Liquid
Refractive index
1.5240 to 1.5270 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text