Thermo Scientific Chemicals

Dimethylvinylchlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: 336730050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethylvinylchlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: 336730050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 336730050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1719-58-0
IUPAC Name
[(1E)-2-chloroethenyl]dimethylsilyl
Molecular Formula
C4H8ClSi
InChI Key
RABBDIBWJVOAPB-ONEGZZNKSA-N
SMILES
C[Si](C)C=CCl
Appearance (Color)
Clear colorless to yellow to amber
Refractive index
1.4140 to 1.4170 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text