4-Nitrophenyl-beta-D-glucuronide, 99+%, Thermo Scientific Chemicals
4-Nitrophenyl-beta-D-glucuronide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl-beta-D-glucuronide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
50 mg
250 mg
Packaging:
Glass bottle
Catalog number 336790010
Price (USD)/ Each
351.65
Online exclusive
391.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
351.65
Online exclusive
391.00 
Save 39.35 (10%)
Add to cart
4-Nitrophenyl-beta-D-glucuronide, 99+%, Thermo Scientific Chemicals
Catalog number336790010
Price (USD)/ Each
351.65
Online exclusive
391.00 
Save 39.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10344-94-2
IUPAC Name(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
Molecular FormulaC12H12NO9
InChI KeyQSUILVWOWLUOEU-GOVZDWNOSA-M
SMILESO[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Water=<8 % (K.F.)
Infrared spectrumConforms
HPLC=<0.05 % (p-Nitrophenol)
Solubility(1 % in water) Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only