Chemical Identifiers
CAS10344-94-2
IUPAC Name(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
Molecular FormulaC12H12NO9
InChI KeyQSUILVWOWLUOEU-GOVZDWNOSA-M
SMILESO[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O
View more