Thermo Scientific Chemicals

α-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%

Catalog number: 337030025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

α-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%

Catalog number: 337030025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 337030025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19046-60-7
IUPAC Name
dipotassium (2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphate
Molecular Formula
C6H11K2O9P
InChI Key
KCIDZIIHRGYJAE-YGFYJFDDSA-L
SMILES
[K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
Appearance (Color)
Colorless to white
Infrared spectrum
Conforms
Specific optical rotation
+81° to +84° (20°C, 589 nm) (c=2, H2O)
Water
=<23 % (K.F.)
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text