This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Specific optical rotation-32.0° to -36.0° (20°C, 589 nm) (c=1, DMF)
UV Lambda max327 to 331 nm (c=1, CH3OH)
Molar extinction coefficient>=17900 l/(mol.cm) (327 to 331 nm, c=1, CH3OH)
UV Lambda max266 to 270 nm (c=1, CH3OH)