Chemical Identifiers
CAS38597-12-5
IUPAC Name4-methyl-7-{[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC16H18O8
InChI KeyYUDPTGPSBJVHCN-HWSQZMMLNA-N
SMILESCC1=CC(=O)OC2=CC(O[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
View more