Thermo Scientific Chemicals

4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 337220050
5 g, Each
Thermo Scientific Chemicals

4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 337220050
5 g, Each
Quantity
Catalog number: 337220050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6160-80-1
IUPAC Name
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxane-2-carboxylic acid
Molecular Formula
C16H16O9
InChI Key
ARQXEQLMMNGFDU-JHZZJYKESA-N
SMILES
CC1=CC(=O)OC2=CC(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)=CC=C12
Appearance (Color)
Colorless to white
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
HPLC
>=97.5 %
Solubility
(c=1, H2O at 60°C) Clear colorless solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text