Creatine phosphate disodium salt tetrahydrate, 99%
Creatine phosphate disodium salt tetrahydrate, 99%
Creatine phosphate disodium salt tetrahydrate, 99%
Creatine phosphate disodium salt tetrahydrate, 99%
Thermo Scientific Chemicals

Creatine phosphate disodium salt tetrahydrate, 99%

CAS: 71519-72-7 | C4H8N3Na2O5P | 255.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
1 g
Packaging:
Glass bottle
Catalog number 337340100
Price (USD)
236.65
Online Exclusive
263.00
Save 26.35 (10%)
Each
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
236.65
Online Exclusive
263.00
Save 26.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS71519-72-7
IUPAC Namedisodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular FormulaC4H16N3Na2O9P
InChI KeyHUWYWJSJJDCZRQ-UHFFFAOYSA-L
SMILESO.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
Water21 to 25 % (K.F.)
Solubility(10 % in water) Clear colorless solution
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection