Thermo Scientific Chemicals

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane, Thermo Scientific Chemicals

Catalog number: 337470050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane, Thermo Scientific Chemicals

Catalog number: 337470050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 337470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
126519-89-9
IUPAC Name
4-[2-(trimethoxysilyl)ethyl]benzene-1-sulfonyl chloride
Molecular Formula
C11H17ClO5SSi
InChI Key
NYIDSUMRGUILGR-UHFFFAOYSA-N
SMILES
CO[Si](CCC1=CC=C(C=C1)S(Cl)(=O)=O)(OC)OC
Infrared spectrum
Conforms
Assay
45 to 55 % solids
Appearance (Color)
Clear yellow to light brown
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text