3,5-Diphenylpyrazole, 99%, Thermo Scientific Chemicals
3,5-Diphenylpyrazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Diphenylpyrazole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 337870050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3,5-diphenyl-1H-pyrazole
Molecular FormulaC15H12N2
InChI KeyJXHKUYQCEJILEI-UHFFFAOYSA-N
SMILESN1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)220.28
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point200°C to 204°C
Titration with HClO4>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only