Thermo Scientific Chemicals

tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%, Thermo Scientific Chemicals

Catalog number: 337880010
1 g, Each
Thermo Scientific Chemicals

tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%, Thermo Scientific Chemicals

Catalog number: 337880010
1 g, Each
Quantity
Catalog number: 337880010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
161671-34-7
IUPAC Name
(1R)-3-(tert-butoxy)-3-oxo-1-phenylpropan-1-aminium
Molecular Formula
C13H20NO2
InChI Key
TYYCBAISLMKLMT-LLVKDONJSA-O
SMILES
CC(C)(C)OC(=O)C[C@@H]([NH3+])C1=CC=CC=C1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration with HClO4
96.0 to 104.0 %
Refractive index
1.4960 to 1.4980 (20°C, 589 nm)
Specific optical rotation
+16° to +21° (20°C, 589 nm) (c=1, CHCl3)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text