Thermo Scientific Chemicals

tert-Butyl (3S)-3-amino-5-methylhexanoate, 95%, Thermo Scientific Chemicals

Catalog number: 337931000
100 mg, Each
Thermo Scientific Chemicals

tert-Butyl (3S)-3-amino-5-methylhexanoate, 95%, Thermo Scientific Chemicals

Catalog number: 337931000
100 mg, Each
Quantity
Catalog number: 337931000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
166023-30-9
IUPAC Name
(3S)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Molecular Formula
C11H24NO2
InChI Key
WBOPVTYXLFEVGP-VIFPVBQESA-O
SMILES
CC(C)C[C@H]([NH3+])CC(=O)OC(C)(C)C
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid or solid
Refractive index
1.4300 to 1.4350 (20°C, 589 nm)
GC
>=94 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text