Thermo Scientific Chemicals

tert-Butyl (3R)-3-aminobutanoate, 95%, Thermo Scientific Chemicals

Catalog number: 337955000
500 mg, Glass Bottle, Each
Thermo Scientific Chemicals

tert-Butyl (3R)-3-aminobutanoate, 95%, Thermo Scientific Chemicals

Catalog number: 337955000
500 mg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 337955000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
158849-23-1
IUPAC Name
(2R)-4-(tert-butoxy)-4-oxobutan-2-aminium
Molecular Formula
C8H18NO2
InChI Key
BFFNZGWJTHWUMY-ZCFIWIBFSA-O
SMILES
C[C@@H]([NH3+])CC(=O)OC(C)(C)C
Refractive index
1.4240 to 1.4260 (20°C, 589 nm)
Specific optical rotation
-21° to -27° (20°C, 589 nm) (C=1, CHCl3)
GC
>=94 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text