Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 95%, Thermo Scientific Chemicals

Catalog number: 338140500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 95%, Thermo Scientific Chemicals

Catalog number: 338140500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 338140500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4039-32-1
IUPAC Name
lithium(1+) bis(trimethylsilyl)azanide
Molecular Formula
C6H18LiNSi2
InChI Key
YNESATAKKCNGOF-UHFFFAOYSA-N
SMILES
[Li+].C[Si](C)(C)[N-][Si](C)(C)C
Appearance (Color)
White to light brown
Appearance (Form)
Crystalline powder and/or chunks
Infrared spectrum
Conforms
Titration with HClO4
94 to 106 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text