3-Methoxy-1,2-propanediol, 98%
3-Methoxy-1,2-propanediol, 98%
3-Methoxy-1,2-propanediol, 98%
3-Methoxy-1,2-propanediol, 98%
Thermo Scientific Chemicals

3-Methoxy-1,2-propanediol, 98%

CAS: 623-39-2 | C4H10O3 | 106.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 338390250
Price (USD)
170.65
Special offer
Online exclusive
201.00
Save 30.35 (15%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
170.65
Special offer
Online exclusive
201.00
Save 30.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS623-39-2
IUPAC Name3-methoxypropane-1,2-diol
Molecular FormulaC4H10O3
InChI KeyPSJBSUHYCGQTHZ-UHFFFAOYNA-N
SMILESCOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4420 to 1.4450 (20°C, 589 nm)
GC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469264Certificate of AnalysisMay 17, 2025338390250, 338390050, 338390010
A0406850Certificate of AnalysisApr 15, 2024338390250, 338390050, 338390010
A0401893Certificate of AnalysisDec 14, 2022338390250, 338390050, 338390010
A0394956Certificate of AnalysisSep 21, 2020338390250, 338390050, 338390010
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection