Nicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals
Nicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 338411000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20260-53-1
IUPAC Namehydrogen pyridine-3-carbonyl chloride chloride
Molecular FormulaC6H5Cl2NO
InChI KeyMSYBLBLAMDYKKZ-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point151°C to 156°C
Titration RCOCl96.0 to 104.0 %
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only