Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals
Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 338700010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(ethenyloxy)trimethylsilane
Molecular FormulaC5H12OSi
InChI KeyHFTNNOZFRQLFQB-UHFFFAOYSA-N
SMILESC[Si](C)(C)OC=C
Molecular Weight (g/mol)116.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.3885 to 1.3905 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only