Thermo Scientific Chemicals

3-(Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: 338930250
Each
Thermo Scientific Chemicals

3-(Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: 338930250
Each
Catalog number: 338930250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17857-14-6
IUPAC Name
(3-carboxypropyl)triphenylphosphanium bromide
Molecular Formula
C22H22BrO2P
InChI Key
NKVJKVMGJABKHV-UHFFFAOYSA-N
SMILES
[Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Appearance (Form)
Powder and/or granules
Melting point
243°C to 250°C
Titration Argentometric
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text