3-(Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
3-(Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Catalog number 338931000
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
3-(Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
Catalog number338931000
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
CAS17857-14-6
IUPAC Name(3-carboxypropyl)triphenylphosphanium bromide
Molecular FormulaC22H22BrO2P
InChI KeyNKVJKVMGJABKHV-UHFFFAOYSA-N
SMILES[Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point243°C to 250°C
Titration Argentometric>=96.0 %
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Powder and/or granules
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only