3-Aminobenzamide, 98%, Thermo Scientific Chemicals
3-Aminobenzamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminobenzamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 339080250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3544-24-9
IUPAC Name3-aminobenzamide
Molecular FormulaC7H8N2O
InChI KeyGSCPDZHWVNUUFI-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point111°C to 115°C
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)Beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only