Thermo Scientific Chemicals

Methyl Red hydrochloride, ACS reagent

Catalog number: 339321000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl Red hydrochloride, ACS reagent

Catalog number: 339321000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 339321000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63451-28-5
IUPAC Name
hydrogen 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid chloride
Molecular Formula
C15H16ClN3O2
InChI Key
ICTWKXAVJAHRMK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
Appearance (Color)
Blue to dark purple
Visual transition interval
From pH 4.2 (pink) to pH 6.2 (yellow)
Appearance (Form)
Powder
Infrared spectrum
Conforms
Clarity of alcohol solution
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text