Chemical Identifiers
CAS20262-76-4
IUPAC Namesodium 4-{[4-(diethylamino)phenyl][4-(diethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonate
Molecular FormulaC27H31N2NaO7S2
InChI KeyPMLFOMWMYRKZRF-UHFFFAOYSA-M
SMILES[Na+].CCN(CC)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC)C1=CC(O)=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
View more