Thermo Scientific Chemicals

Patent Blue V, sodium salt, Thermo Scientific Chemicals

Catalog number: 339331000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Patent Blue V, sodium salt, Thermo Scientific Chemicals

Catalog number: 339331000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 339331000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20262-76-4
IUPAC Name
sodium 4-{[4-(diethylamino)phenyl][4-(diethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonate
Molecular Formula
C27H31N2NaO7S2
InChI Key
PMLFOMWMYRKZRF-UHFFFAOYSA-M
SMILES
[Na+].CCN(CC)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC)C1=CC(O)=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
Appearance (Color)
Dark blue-green
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Dye content
>=80 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text