Thermo Scientific Chemicals

tert.-Butyl methyl malonate, 95%, Thermo Scientific Chemicals

Catalog number: 339390250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

tert.-Butyl methyl malonate, 95%, Thermo Scientific Chemicals

Catalog number: 339390250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 339390250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
42726-73-8
IUPAC Name
1-tert-butyl 3-methyl propanedioate
Molecular Formula
C8H14O4
InChI Key
XPSYZCWYRWHVCC-UHFFFAOYSA-N
SMILES
COC(=O)CC(=O)OC(C)(C)C
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Appearance
May discolor to yellow on storage
Infrared spectrum
Conforms
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text