Thermo Scientific Chemicals

2-Methyl-DL-serine hydrate, 96%

Catalog number: 339750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methyl-DL-serine hydrate, 96%

Catalog number: 339750050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 339750050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5424-29-3
IUPAC Name
2-amino-3-hydroxy-2-methylpropanoic acid
Molecular Formula
C4H9NO3
InChI Key
CDUUKBXTEOFITR-UHFFFAOYNA-N
SMILES
CC(N)(CO)C(O)=O
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Titration with HClO4
>=95 % (On dry substance)
Infrared spectrum
Conforms
Water
=<10 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text