Thermo Scientific Chemicals

DL-2-Benzylserine, 97%

Catalog number: 339780100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-2-Benzylserine, 97%

Catalog number: 339780100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 339780100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4740-47-0
IUPAC Name
(2S)-2-azaniumyl-2-benzyl-3-hydroxypropanoate
Molecular Formula
C10H13NO3
InChI Key
ZMNNAJIBOJDHAF-JTQLQIEISA-N
SMILES
[NH3+][C@](CO)(CC1=CC=CC=C1)C([O-])=O
Appearance (Color)
Beige to light brown
Appearance (Form)
Crystalline powder or flakes
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text