1x1 image pixel for data collection
ONPG (o-nitrophenyl-β-D-galactopyranoside)
ONPG (o-nitrophenyl-β-D-galactopyranoside)
Thermo Scientific™
ONPG (o-nitrophenyl-β-D-galactopyranoside)
Thermo Scientific Pierce ONPG is the preferred colorimetric substrate for ELISA applications involving beta-galactosidase (b-Gal) as the reporter enzyme.When usingRead more
Have Questions?
Catalog NumberQuantity
340555 g
Catalog number 34055
Price (USD)
135.00
Each
In stock
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
135.00
Each
Add to cart
Thermo Scientific Pierce ONPG is the preferred colorimetric substrate for ELISA applications involving beta-galactosidase (b-Gal) as the reporter enzyme.
When using beta-Galactosidase as the label for proteins in ELISA studies, a wide variety of substrates are available, including: o-Nitrophenyl-beta-D-galactopyranosidase (ONPG), Naphthol-AS-Bl-beta-D-galactopyranosidase (Nap-Gal), and 4-Methyl-umbelliferyl-beta-D-galactopyranosidase (MUm-Gal). However, it is important to choose a substrate with adequate solubility, that uses readily available equipment, and that gives a significant reading over the background. ONPG is a superior beta-Galactosidase substrate option.(1) The product formed is completely soluble and has a high extinction coefficient at 405nm. The substrate yields a yellow product that is easily detectable in the visual range after stopping the reaction with 1M Sodium Carbonate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodColorimetric
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Gal
Product TypeELISA Substrate
Quantity5 g
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C protected from moisture and light. Product is shipped at ambient temperature.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link