Thermo Scientific™

Pierce™ Diethanolamine Substrate Buffer (5X)

Catalog number: 34064
Have Questions?
Thermo Scientific™

Pierce™ Diethanolamine Substrate Buffer (5X)

Catalog number: 34064
Have Questions?
Catalog Number
34064
Unit Size
225 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
34064225 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Diethanolamine Substrate Buffer (5X) include optimized, reliable and convenient stock solutions of diethanolamine for alkaline phosphatase (alk-phos) detection.

Features of Diethanolamine Substrate Buffer (5X):

• Ready-to-use
• Minimize assay-to-assay variability

Diethanolamine Substrate Buffer is used with soluble alkaline phosphatase substrates such as PNPP. Our formulation is convenient, ready-to-use and reduces the possibility of assay-to-assay variability. Diethanolamine Substrate Buffer is sold as 5X concentrate with an optimal, consistent pH. It is stable even at a 1X concentration.

Related Products
Pierce™ Stable Peroxide Substrate Buffer (10X)
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Pierce Diethanolamine Substrate Buffer, 5X
Product Type
ELISA Substrate
Form
Liquid
For Use With (Application)
ELISA, Immunohistochemistry (IHC), Western Blot
Product Line
Pierce™
Substrate Type
Alkaline Phosphatase, Horseradish Peroxidase

Contents & Storage

Upon receipt store product at 4°C. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text