Thermo Scientific™

1-Step™ NBT/BCIP plus Suppressor Substrate Solution

Catalog number: 34070
Have Questions?
Thermo Scientific™

1-Step™ NBT/BCIP plus Suppressor Substrate Solution

Catalog number: 34070
Have Questions?
Catalog Number
34070
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
34070100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).

Features:

AP substrate—for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
Chromogenic—no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph
Package options—choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor

Highlights of Pierce NBT/BCIP 1-Step Solution (Part No. 34042):
• Best for immunoblotting applications
• Ready-to-use single component
• Sensitive with low background and minimial assay-to-assay variability

Highlights of Pierce NBT/BCIP plus Suppressor 1-Step Solution (Part No. 34070):
• Best for immunohistochemistry applications
• Ready-to-use single component
• Contains 1 mM levamisole to inhibit endogenous phosphatase activity, while yielding an intense dark purple precipitate in the presence of calf intestinal phosphatase

Related Products
1-Step™ NBT/BCIP Substrate Solution
NBT Substrate Powder (nitro-blue tetrazolium chloride)
BCIP Substrate Powder (5-bromo-4-chloro-3'-indolyphosphate p-toluidine)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Pierce 1-Step NBT/BCIP Plus Suppressor Substrate Solution
Product Type
Substrate and Suppressor
Detection Method
Colorimetric
Form
Liquid
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
1-Step™
Quantity
100 mL
Substrate
Alkaline Phosphatase
Reactivity
Alkaline Phosphatase

Contents & Storage

Upon receipt store at 4°C. Product is shipped with an ice pack.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text