Thermo Scientific™

SuperSignal™ West Femto Maximum Sensitivity Substrate

Catalog number: 34094
Have Questions?
Thermo Scientific™

SuperSignal™ West Femto Maximum Sensitivity Substrate

Catalog number: 34094
Have Questions?
Catalog Number
34094
Unit Size
20 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 200 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
3409420 mL
-
34095X44 x 100 mL
-
34095100 mL
-
34096X44 x 200 mL
-
34096200 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific SuperSignal West Femto Maximum Sensitivity Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for low femtogram protein level detection by western blot analysis with horseradish peroxidase (HRP) enzyme.

SuperSignal West Femto Maximum Sensitivity Substrate characteristics include:
Sensitivity: low-femtogram (mid-zeptomole)
Stability: 8-hr working solution stability, 1-year kit stability at 4°C
Compatibility: nitrocellulose and PVDF membranes
Signal duration: up to 8 hours
Recommended primary antibody concentration: 1:5,000–1:100,000 dilution (10–200 ng/mL)
Recommended secondary antibody concentration: 1:100,000–1:500,000 dilution (2–10 ng/mL)

Compare and learn more about our western blot chemiluminescent substrates ›
View all HRP substrates available ›

SuperSignal West Femto Maximum Sensitivity Substrate is designed for low femtogram-level detection by western blot analysis. When combined with optimized antibody concentrations and blocking buffers, SuperSignal West Femto substrate enables detection of target proteins in amounts that are too small to be seen with typical ECL substrates.

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Substrate
Detection Method
Chemiluminescent
Form
Liquid
For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
SuperSignal™
Quantity
20 mL
Substrate
Horseradish Peroxidase
Recommended Antibody Concentrations
2°: 1:100-500K (2-10ng/mL), 1°: 1:5K (10- 200ng/mL)
Sensitivity
Low femtogram-level
Signal Duration
6 to 8 hr.
Sufficient For
2 Mini-Gel Blots, 200 sq. cm of Membrane
Kit Contents
Stable Peroxide Buffer, 10mL, Luminol/Enhancer, 10mL

Contents & Storage

Sufficient For: 2 mini-gel blots; 200 cm2 of membrane
• Luminol/Enhancer, 10 mL
• Stable Peroxide Buffer, 10 mL

Store reagents at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text