Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Actual product may vary
Thermo Scientific™

Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Closure Size
342032-00055 mL20-415
342032-001010 mL20-415
342032-002020 mL20-415
3 Options
Catalog number 342032-0005
Price (USD)/ Case of 1656
604.00
Online exclusive
755.00 
Save 151.00 (20%)
-
Add to cart
Volume (Metric):
5 mL
Closure Size:
20-415
Price (USD)/ Case of 1656
604.00
Online exclusive
755.00 
Save 151.00 (20%)
Add to cart
Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Catalog number342032-0005
Price (USD)/ Case of 1656
604.00
Online exclusive
755.00 
Save 151.00 (20%)
-
Add to cart

Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).

  • PETG
  • Non-cytotoxic
  • Non-pyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL
  • Leakproof† with Nalgene lined continuous thread closures (Cat. No. 342158)

Ordering Information:

Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
DescriptionSerum Vials
Diameter (Metric) Inner11 mm
ColorClear
Diameter (Metric) Outer22 mm
MaterialPETG
SterilitySterile
Volume (Metric)5 mL
No. per Case1656
No. per Pack276
Volume (English)1/6 oz.
Product LineNalgene
Volume (Metric) Total5 mL
Closure TypeLined Continuous-thread Closure (Sold Separately)
Height (Metric)44 mm
Quantity1656 Cases, 276 Pk.
Certifications/ComplianceISO 13485:2016
TypeSerum Vial
Closure Size20-415
For Use With (Equipment)Lined Continuous-Thread Closure
Unit SizeCase of 1656