Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Combine the elastomeric sealing of silicone with nonreactive polytetrafluoroethylene reagent contact surfaces in Thermo Scientific™ Nalgene™ Septum Closures, for use with Nalgene™ Sterile PET & PETG media bottles.
Color | White |
Description | Nalgene Closure for Media Bottles |
Material | HDPE |
No. per Case | 200 |
No. per Pack | 200 |
Packaging | Bulk |
Sterility | Sterile |
Liner Material | Silicone, PTFE |
Certifications/Compliance | Manufactured in compliance with ISO 13485 quality requirements |
Closure Included | Yes |
Fit for Purpose | Sterile |
Product Type | Closure |
For Use With (Equipment) | Nalgene Sterile PET and PETG media bottles |
Closure Material | Polyethylene |
Closure Size | 24-415 mm |
Unit Size | Case of 200 |
Color | White |
Description | Nalgene Closure for Media Bottles |
Material | HDPE |
No. per Case | 240 |
No. per Pack | 240 |
Packaging | Bulk |
Sterility | Sterile |
Liner Material | Silicone, PTFE |
Certifications/Compliance | Manufactured in compliance with ISO 13485 quality requirements |
Closure Included | Yes |
Fit for Purpose | Sterile |
Product Type | Closure |
For Use With (Equipment) | Nalgene Sterile PET and PETG media bottles |
Closure Material | Polyethylene |
Closure Size | 38-430 mm |
Unit Size | Case of 240 |
Catalog Number | Specifications | Unit Size | dimClosureSize | No. per Case | Fit for Purpose | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
342178-0240 | Case of 200 | 200 | Sterile | 548.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
342178-0384 | Case of 240 | 240 | Sterile | 697.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Note:
Bottles are tested during each manufacturing run to ensure leakproof† performance. Suitable for air shipment. †The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.