Thermo Scientific Chemicals

Sodium perchlorate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 342182500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium perchlorate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 342182500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 342182500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7601-89-0
IUPAC Name
sodium perchlorate
Molecular Formula
ClNaO4
InChI Key
BAZAXWOYCMUHIX-UHFFFAOYSA-M
SMILES
[Na+].[O-][Cl](=O)(=O)=O
Appearance (Color)
White
Appearance (Form)
Powder or crystals and/or chunks
Titration after Ion exchange
97.5 to 102.5 %
pH
6.0 to 8.0 (5% in water, 25°C)
Insoluble matter in water
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text