Thermo Scientific Chemicals

6-Hydroxycaproic acid, 95%, Thermo Scientific Chemicals

Catalog number: 342240050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Hydroxycaproic acid, 95%, Thermo Scientific Chemicals

Catalog number: 342240050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 342240050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1191-25-9
IUPAC Name
6-hydroxyhexanoate
Molecular Formula
C6H11O3
InChI Key
IWHLYPDWHHPVAA-UHFFFAOYSA-M
SMILES
OCCCCCC([O-])=O
Appearance (Color)
White
Appearance (Form)
Low melting crystalline solid
Infrared spectrum
Conforms
GC
>=94 %
Additional info
may contain variable amounts of dimers

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text