Thermo Scientific Chemicals

4-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 342460050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 342460050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 342460050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16004-15-2
IUPAC Name
1-(bromomethyl)-4-iodobenzene
Molecular Formula
C7H6BrI
InChI Key
BQTRMYJYYNQQGK-UHFFFAOYSA-N
SMILES
BrCC1=CC=C(I)C=C1
Appearance (Color)
White to light yellow
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
GC
>=96.0 %
Melting point
77°C to 82°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text