Formononetin, 98%, Thermo Scientific Chemicals
Formononetin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Formononetin, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 342480010
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
167.00 
Save 25.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
167.00 
Save 25.35 (15%)
Add to cart
Formononetin, 98%, Thermo Scientific Chemicals
Catalog number342480010
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
167.00 
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
CAS485-72-3
IUPAC Name7-hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
Molecular FormulaC16H12O4
InChI KeyHKQYGTCOTHHOMP-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point256°C to 261°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only