Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Thermo Scientific™

Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack

Protect light sensitive reagents with these PPCO amber micro packaging vials, molded from high purity, noncytotoxic, nonpyrogenic, low-metal content polypropylene copolymer (PPCO) resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
342805-00202 mL
342805-00050.5 mL
Catalog number 342805-0020
Price (MXN)
-
Volume (Metric):
2 mL
Ask our AI about this Product
Protect light sensitive reagents with Thermo Scientific™ Nalgene™ PPCO Amber Micro Packaging Vials. These noncytotoxic and nonpyrogenic vials are molded from sterile high purity, low-metal content polypropylene copolymer resin and are ready to use. Vials are skirted and feature a conical bottom, allowing recovery of entire contents.
Features:
  • Vials and closures (listed separately) pressure-tested together at 7.5 PSIG
  • USP Class VI compliant; non-pyrogenic
  • Can be centrifuged at 13,000xg
  • Components provided sterile and ready to fill
  • Colored closures packaged separately
  • Sterile to 10-6 SAL
  • RNase/DNase free
  • Are leakproof† when used with Nalgene closures

Ordering Information:

Micro Packaging Vials are bulk packed without closures, 500 per inner bag, 1,000 per case. Order sterile closures separately (Cat. Nos. 342821, 342825).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Closure MaterialPPCO
Closure Size11 mm
DescriptionMicro Packaging Vials
Diameter (Metric) Inner8 mm
No. per Case1000
No. per Pack500
StyleSkirted vial with conical bottom
Volume (Metric) Total2 mL
Certifications/ComplianceISO 13485:2016
Closure TypeScrew Cap (Not Included)
ColorAmber
Diameter (Metric) Outer13 mm
Height (Metric)49 mm
MaterialPPCO
Quantity500 Bag, 1000 Cs.
SterilitySterile
TypeVial
Volume (Metric)2 mL
Unit SizeCase of 1000
nalgene-certificate-of-compliance

Certificates of Compliance

Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Nalgene and Nunc products.
Start your request  

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link