Captopril, 98%
Thermo Scientific Chemicals

Captopril, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 343700050
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
Add to cart
Captopril, 98%
Catalog number343700050
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
Molecular FormulaC9H15NO3S
InChI KeyFAKRSMQSSFJEIM-RQJHMYQMSA-N
SMILESC[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O
Molecular Weight (g/mol)217.28
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point103°C to 108°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only