Thermo Scientific Chemicals

Captopril, 98%

Catalog number: 343700250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Captopril, 98%

Catalog number: 343700250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 343700250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62571-86-2
IUPAC Name
(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
Molecular Formula
C9H15NO3S
InChI Key
FAKRSMQSSFJEIM-RQJHMYQMSA-N
SMILES
C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
103°C to 108°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text