Thermo Scientific Chemicals

Sodium phosphate dibasic dihydrate, 99+%, for HPLC, Thermo Scientific Chemicals

Catalog number: 343810025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Sodium phosphate dibasic dihydrate, 99+%, for HPLC, Thermo Scientific Chemicals

Catalog number: 343810025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 343810025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10028-24-7
IUPAC Name
disodium dihydrate hydrogen phosphate
Molecular Formula
H5Na2O6P
InChI Key
KDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILES
O.O.[Na+].[Na+].OP([O-])([O-])=O
Appearance (Color)
White
pH
7.5 to 9.2 (5 % at 25°C)
Appearance (Form)
Crystalline powder or crystals
Lead (Pb)
=<5 ppm
UV
(0.5 M in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text