Sodium phosphate dibasic dihydrate, 99+%, for HPLC, Thermo Scientific Chemicals
Sodium phosphate dibasic dihydrate, 99+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium phosphate dibasic dihydrate, 99+%, for HPLC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 343811000
Price (USD)/ Each
48.65
Online exclusive
56.90 
Save 8.25 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
56.90 
Save 8.25 (15%)
Add to cart
Sodium phosphate dibasic dihydrate, 99+%, for HPLC, Thermo Scientific Chemicals
Catalog number343811000
Price (USD)/ Each
48.65
Online exclusive
56.90 
Save 8.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium dihydrate hydrogen phosphate
Molecular FormulaH5Na2O6P
InChI KeyKDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OP([O-])([O-])=O
Molecular Weight (g/mol)177.99
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH7.5 to 9.2 (5 % at 25°C)
Appearance (Form)Crystalline powder or crystals
Lead (Pb)=<5 ppm
UV(0.5 M in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only