Thermo Scientific Chemicals

Tetramethylammonium hydrogensulfate, 99+%, HPLC grade, Thermo Scientific Chemicals

Catalog number: 343830500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetramethylammonium hydrogensulfate, 99+%, HPLC grade, Thermo Scientific Chemicals

Catalog number: 343830500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 343830500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80526-82-5
IUPAC Name
tetramethylazanium hydrogen sulfate
Molecular Formula
C4H13NO4S
InChI Key
DWTYPCUOWWOADE-UHFFFAOYSA-M
SMILES
C[N+](C)(C)C.OS([O-])(=O)=O
Appearance (Form)
Powder
Water
=<1 % (K.F.)
UV
at 220 nm A: =<0.05
UV
at 200 nm A: =<0.15
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text