Thermo Scientific Chemicals

L(-)-Glutathione, oxidized hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 343840010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

L(-)-Glutathione, oxidized hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 343840010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 343840010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27025-41-8
IUPAC Name
(2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid
Molecular Formula
C20H32N6O12S2
InChI Key
YPZRWBKMTBYPTK-IOKZUGQQNA-N
SMILES
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Water
=<8 % (K.F.)
Sulfated ash
=<0.1 % (800°C)
Iron (Fe)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text