Thermo Scientific Chemicals

Brij™ C20, Thermo Scientific Chemicals

Catalog number: 344290025
2.5 kg, Each
Thermo Scientific Chemicals

Brij™ C20, Thermo Scientific Chemicals

Catalog number: 344290025
2.5 kg, Each
Quantity
Catalog number: 344290025
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
9004-95-9
MDL Number
MFCD00080892
Appearance (Color)
White
Water
=<1 %
Impurity
=<1 ppm (Free ethyleneoxide)
Appearance (Form)
Pellets
Hydroxyl value
45 to 60 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text