Carbamazepine, 98%
Thermo Scientific Chemicals

Carbamazepine, 98%

Carbamazepine (Carbamazepen), CAS # 298-46-4, is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker.
Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 344360250
Price (USD)/ Each
186.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
219.00 
Save 32.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
186.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
219.00 
Save 32.35 (15%)
Add to cart
Carbamazepine, 98%
Catalog number344360250
Price (USD)/ Each
186.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
219.00 
Save 32.35 (15%)
-
Add to cart
Chemical Identifiers
CAS298-46-4
IUPAC Name2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
Molecular FormulaC15H12N2O
InChI KeyFFGPTBGBLSHEPO-UHFFFAOYSA-N
SMILESNC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=97.5 %
Appearance (Form)Powder or crystals
Melting point189°C to 193°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Carbamazepine is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker
• It is a tricyclic antidepressant-like drug that reduces post-tetanic potentiation
Applications
Carbamazepine can be used as a sodium channel blocker and as a glutamate-transporter agonist to reduce neural activity and produce inhibitory effects in the brain
Carbamazepine is useful in the research of pain, epilepsy, and manic depression
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Ben-Menachem, E.; Grebe, H.P.; Terada, K.; Jensen, L.; Li, T.; De Backer, M.; Steiniger-Brach, B.; Gasalla, T.; Brock, M.; Biton, V. Long-term safety and efficacy of lacosamide and controlled-release carbamazepine monotherapy in patients with newly diagnosed epilepsy. Epilepsia. 2019, 60(12), 2437-2447. Erratum in: Epilepsia. 2021, 62(4), 1039.
  2. Schmitz, B.; Dimova, S.; Zhang, Y.; Chellun, D.; De Backer, M.; Gasalla, T. Tolerability and efficacy of lacosamide and controlled-release carbamazepine monotherapy in patients with newly diagnosed epilepsy and concomitant psychiatric conditions: Post hoc analysis of a prospective, randomized, double-blind trial. Epilepsy Res. 2020, 159, 106220.