Thermo Scientific Chemicals

N-tert-Butoxycarbonyl-L-prolinol, 98%, Thermo Scientific Chemicals

Catalog number: 344560250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N-tert-Butoxycarbonyl-L-prolinol, 98%, Thermo Scientific Chemicals

Catalog number: 344560250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 344560250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
69610-40-8
IUPAC Name
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular Formula
C10H19NO3
InChI Key
BFFLLBPMZCIGRM-QMMMGPOBSA-N
SMILES
CC(C)(C)OC(=O)N1CCC[C@H]1CO
Melting point
58°C to 63°C
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Specific optical rotation
-46° to -52° (20°C, 589 nm) (c=1, CHCl3)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text