D(+)-Phenyllactic acid, 98%, Thermo Scientific Chemicals
D(+)-Phenyllactic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D(+)-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 344580050
Price (USD)/ Each
172.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
203.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
172.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
203.00 
Save 30.35 (15%)
Add to cart
D(+)-Phenyllactic acid, 98%, Thermo Scientific Chemicals
Catalog number344580050
Price (USD)/ Each
172.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
203.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7326-19-4
IUPAC Name(2R)-2-hydroxy-3-phenylpropanoic acid
Molecular FormulaC9H10O3
InChI KeyVOXXWSYKYCBWHO-MRVPVSSYSA-N
SMILESO[C@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point121°C to 125°C
Titration with NaOH>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only