Thermo Scientific Chemicals

Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals

Catalog number: 344590500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals

Catalog number: 344590500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 344590500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10543-57-4
IUPAC Name
N-acetyl-N-[2-(N-acetylacetamido)ethyl]acetamide
Molecular Formula
C10H16N2O4
InChI Key
BGRWYDHXPHLNKA-UHFFFAOYSA-N
SMILES
CC(=O)N(CCN(C(C)=O)C(C)=O)C(C)=O
Appearance (Form)
Granular powder
Infrared spectrum
Conforms
Assay
>=88 %
Appearance (Color)
Off-white to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text