Thermo Scientific Chemicals

3-Carboxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 344630050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Carboxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 344630050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 344630050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25487-66-5
IUPAC Name
3-(dihydroxyboranyl)benzoic acid
Molecular Formula
C7H7BO4
InChI Key
DBVFWZMQJQMJCB-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC(=C1)C(O)=O
Appearance (Color)
Off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text